3-Bromo-2,5-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A359332-100MG A359332-250MG A359332-1G
77015-846EA 38.2 USD
77015-846 77015-848 77613-048
3-Bromo-2,5-difluorobenzaldehyde 98%
3-Bromo-2,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
MDL Number: MFCD26401549
CAS Number: 112279-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR