(2-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A359196-1G
A359196-250MG
A359196-5G
77015-840EA
28.99
USD
77015-840
77015-842
77015-844
(2-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 98%
(2-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₉BO₄ |
MDL Number:
MFCD07784402 CAS Number: 871126-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...