1-(4-Bromophenyl)cyclopropanecarbonitrile 97%

Supplier: Ambeed
Warning

A358741-1G A358741-25G A358741-5G A358741-100G A358741-10G
76989-784EA 9 USD
76989-784 76989-786 76989-788 77885-947 77885-948
1-(4-Bromophenyl)cyclopropanecarbonitrile 97%
1-(4-Bromophenyl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD01314308
CAS Number: 124276-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR