4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A358689-100MG A358689-250MG
77015-836EA 28.65 USD
77015-836 77015-838
4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
4,4-Dimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClN
MW: 197.7 g/mol
MDL Number: MFCD11501295
CAS Number: 41565-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR