5-Nitro-3H-benzofuran-2-one 97%

Supplier: Ambeed
Warning

Synonyms: 5-Nitrobenzofuran-2(3H)-one

A358484-1G A358484-250MG
77033-166EA 94.43 USD
77033-166 77033-168
5-Nitro-3H-benzofuran-2-one 97%
5-Nitro-3H-benzofuran-2-one

Precaución: for Research Use Only

Formula: C₈H₅NO₄
MW: 179.13 g/mol
MDL Number: MFCD18071597
CAS Number: 21997-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR