3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride 96%, Ambeed.Inc
Supplier: Ambeed
|
Danger
|
A358480-100MG
A358480-1G
A358480-250MG
A358480-5G
A358480-25G
76989-776EA
13.8
USD
76989-776
76989-778
76989-780
76989-782
77684-054
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride 96%, Ambeed.Inc
3-(Chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride
Precaución: For research use only.
Formula:
C₄H₇Cl₂N₃ |
MDL Number:
MFCD09701417 CAS Number: 135206-76-7 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...