5-Nitro-2-indanone 97%

Supplier: Ambeed
Warning

A358233-1G A358233-250MG A358233-5G
77015-818EA 19 USD
77015-818 77015-820 77015-822
5-Nitro-2-indanone 97%
5-Nitro-2-indanone

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MDL Number: MFCD00598938
CAS Number: 116530-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR