5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride 95%

Supplier: Ambeed
Warning

A358073-10G A358073-250MG A358073-25G
77015-804EA 627 USD
77015-804 77015-806 77015-808
5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride 95%
5,6,7,8-Tetrahydroquinolin-8-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄Cl₂N₂
MW: 221.13 g/mol
MDL Number: MFCD11506266
CAS Number: 1187929-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR