6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A358025-100MG A358025-1G A358025-250MG
77015-798EA 60.48 USD
77015-798 77015-800 77015-802
6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 97%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MW: 177.59 g/mol
MDL Number: MFCD18380875
CAS Number: 1260385-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR