2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Phenylthiazol-4-yl)acetic acid

A357403-100MG A357403-1G
77015-794EA 56.23 USD
77015-794 77015-796
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid 97%
2-(2-Phenyl-1,3-thiazol-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂S
MW: 219.26 g/mol
MDL Number: MFCD01312954
CAS Number: 16441-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR