6-(2,2,2-Trifluoroethoxy)nicotinic acid 98%
Supplier: Ambeed
|
Warning
|
A357365-10G
A357365-1G
A357365-250MG
A357365-5G
77015-786EA
134
USD
77015-786
77015-788
77015-790
77015-792
6-(2,2,2-Trifluoroethoxy)nicotinic acid 98%
6-(2,2,2-Trifluoroethoxy)nicotinic acid
Precaución: for Research Use Only
Formula:
C₈H₆F₃NO₃ |
MDL Number:
MFCD00110708 CAS Number: 175204-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...