(3-Butoxy-2-chlorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

A357334-1G A357334-250MG A357334-5G
77015-780EA 143 USD
77015-780 77015-782 77015-784
(3-Butoxy-2-chlorophenyl)boronic acid 97%
(3-Butoxy-2-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₃
MDL Number: MFCD17015727
CAS Number: 1256345-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR