4-Butylaniline 97%

Supplier: Ambeed
Warning

A356830-25G A356830-500G A356830-100G
77033-154EA 20 USD
77033-154 77033-156 77672-790
4-Butylaniline 97%
4-Butylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₅N
MDL Number: MFCD00007925
CAS Number: 104-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR