N-(2-Hydroxyethyl) 4-boronobenzenesulfonamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-[N-(2-Hydroxyethyl)sulphamoyl]benzeneboronic acid, (4-(N-(2-Hydroxyethyl)sulphamoyl)phenyl)boronic acid
A356714-100MG
A356714-1G
A356714-250MG
76989-752EA
10.61
USD
76989-752
76989-754
76989-756
N-(2-Hydroxyethyl) 4-boronobenzenesulfonamide 98%
N-(2-Hydroxyethyl) 4-boronobenzenesulphonamide
Precaución: for Research Use Only
Formula:
C₈H₁₂BNO₅S MW: 245.0600 g/mol |
MDL Number:
MFCD07363750 CAS Number: 850568-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...