6-Methylbenzo[b]thiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Carboxy-6-methyl-1-benzothiophene, 6-Methyl-1-benzothiophene-2-carboxylic acid

A356711-1G A356711-5G
76989-748EA 43.5 USD
76989-748 76989-750
6-Methylbenzo[b]thiophene-2-carboxylic acid 97%
6-Methylbenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MW: 192.234 g/mol
MDL Number: MFCD07377056
CAS Number: 1467-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR