5-Chloro-3-phenylisoxazole 97%

Supplier: Ambeed
Warning

A356506-100MG A356506-250MG A356506-1G
77015-772EA 136.86 USD
77015-772 77613-026 77684-049
5-Chloro-3-phenylisoxazole 97%
5-Chloro-3-phenylisoxazole

Precaución: For research use only.

Formula: C₉H₆ClNO
MW: 179.60 g/mol
MDL Number: MFCD00046079
CAS Number: 3356-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR