1-Boc-4-(2-aminophenyl)piperazine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Aminophenyl)piperazine, N1-BOC protected, 1-Boc-4-(2-Aminophenyl)piperazine

A355702-1G A355702-250MG A355702-5G
77015-762EA 23 USD
77015-762 77015-764 77015-766
1-Boc-4-(2-aminophenyl)piperazine 97%
1-Boc-4-(2-aminophenyl)piperazine

Precaución: for Research Use Only

Formula: C₁₅H₂₃N₃O₂
MDL Number: MFCD04115046
CAS Number: 170017-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR