(4-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A355267-1G A355267-250MG A355267-5G
77015-756EA 122.01 USD
77015-756 77015-758 77015-760
(4-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid 98%
(4-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₃O₄
MW: 312.05 g/mol
MDL Number: MFCD08276770
CAS Number: 870778-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR