L-Selenomethionine 98%

Supplier: Ambeed
Danger

Synonyms: L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH

A355129-1MG A355129-250MG A355129-1G A355129-25G A355129-100MG
76989-726EA 5.31 USD
76989-726 76989-728 77568-718 77594-054 77613-020
L-Selenomethionine 98%
L-Selenomethionine

Precaución: For research use only.

Formula: C₅H₁₁NO₂Se
MW: 196.11 g/mol
MDL Number: MFCD00037210
CAS Number: 3211-76-5
UN: 3283
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR