[1-(Aminomethyl)cyclopropyl]methanol 98%

Supplier: Ambeed
Danger

Synonyms: (1-(Aminomethyl)cyclopropyl)methanol

A355027-1G A355027-250MG A355027-5G
77033-142EA 128.37 USD
77033-142 77033-144 77033-146
[1-(Aminomethyl)cyclopropyl]methanol 98%
[1-(Aminomethyl)cyclopropyl]methanol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD09864388
CAS Number: 45434-02-4
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR