4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene 97%
Supplier: Ambeed
|
Warning
|
A354942-100MG
A354942-1G
A354942-250MG
A354942-5G
76989-718EA
13
USD
76989-718
76989-720
76989-722
76989-724
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene 97%
4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene
Precaución: for Research Use Only
Formula:
C₂₆H₃₈O₂S₂ |
MDL Number:
MFCD17018551 CAS Number: 1160823-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...