2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylthio)oxazolo[4,5-b]pyridine

A354637-100MG A354637-1G A354637-250MG A354637-5G
76989-704EA 9.55 USD
76989-704 76989-706 76989-708 76989-710
2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine 95%
2-(Methylsulfanyl)[1,3]oxazolo[4,5-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆N₂OS
MW: 166.2 g/mol
MDL Number: MFCD00973738
CAS Number: 169205-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR