(2-Amino-4-chlorophenyl)(phenyl)methanone 95%

Supplier: Ambeed
Warning

A354534-1G A354534-25G A354534-5G A354534-100G
77003-302EA 28 USD
77003-302 77003-304 77003-306 77881-969
(2-Amino-4-chlorophenyl)(phenyl)methanone 95%
(2-Amino-4-chlorophenyl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD00179566
CAS Number: 4076-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR