1-Benzyl-3,3-difluoropyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A354420-100MG A354420-250MG
77015-750EA 45 USD
77015-750 77015-752
1-Benzyl-3,3-difluoropyrrolidine hydrochloride 95%
1-Benzyl-3,3-difluoropyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClF₂N
MW: 233.69 g/mol
MDL Number: MFCD14706680
CAS Number: 862416-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR