Saxagliptin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3S,5S)-2-((2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetyl)-2-azabicyclo[3.1.0]hexane-3-carbonitrile

A354223-10MG A354223-1MG A354223-5MG
77033-126EA 13.8 USD
77033-126 77033-128 77033-130
Saxagliptin 98%
Saxagliptin

Precaución: for Research Use Only

Formula: C₁₈H₂₅N₃O₂
MW: 315.42 g/mol
MDL Number: MFCD12756398
CAS Number: 361442-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR