Methyl-2-(methylsulfonyl)benzenecarboxylate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl 2-(methylsulphonyl)benzoate

A354206-10G A354206-1G
77003-294EA 1159.57 USD
77003-294 77003-296
Methyl-2-(methylsulfonyl)benzenecarboxylate 95%
Methyl-2-(methylsulfonyl)benzenecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
MDL Number: MFCD03001211
CAS Number: 199657-02-8
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR