2-(2-Methoxyphenoxy)-2-methylpropanoic acid 95+%

Supplier: Ambeed
Warning

A353966-1G A353966-250MG
77033-122EA 63.66 USD
77033-122 77033-124
2-(2-Methoxyphenoxy)-2-methylpropanoic acid 95+%
2-(2-Methoxyphenoxy)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MDL Number: MFCD00995567
CAS Number: 53498-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR