2,6-Dichloro-3-pyridinemethanol 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-3-(hydroxymethyl)pyridine, (2,6-Dichloropyridin-3-yl)methanol

A353577-1G A353577-250MG
77003-290EA 37.14 USD
77003-290 77003-292
2,6-Dichloro-3-pyridinemethanol 95%
2,6-Dichloro-3-pyridinemethanol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.012 g/mol
MDL Number: MFCD11036367
CAS Number: 55304-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR