1,5-Difluoro-2-methyl-4-nitrobenzene 96%

Supplier: Ambeed
Warning

A353497-100MG A353497-1G A353497-250MG A353497-5G
76989-682EA 6 USD
76989-682 76989-684 76989-686 76989-688
1,5-Difluoro-2-methyl-4-nitrobenzene 96%
1,5-Difluoro-2-methyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD16620288
CAS Number: 179011-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR