4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95%

Supplier: Ambeed
Danger

Synonyms: 4,7-Bis(5-bromothiophen-2-yl)benzo[c][1,2,5]thiadiazole

A353228-10G A353228-1G A353228-5G
76989-674EA 974.97 USD
76989-674 76989-676 76989-678
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole 95%
4,7-Bis(5-bromo-2-thienyl)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₁₄H₆Br₂N₂S₃
MW: 458.20 g/mol
MDL Number: MFCD16619295
CAS Number: 288071-87-4
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR