5-Hydroxy-4-methoxy-2-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A353204-100MG A353204-1G A353204-250MG A353204-5G
76989-666EA 23 USD
76989-666 76989-668 76989-670 76989-672
5-Hydroxy-4-methoxy-2-nitrobenzaldehyde 95%
5-Hydroxy-4-methoxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MDL Number: MFCD00228444
CAS Number: 58749-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR