tert-Butyl (5-methylthiophen-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A352959-100MG A352959-250MG
76989-662EA 6.37 USD
76989-662 76989-664
tert-Butyl (5-methylthiophen-3-yl)carbamate 97%
tert-Butyl (5-methylthiophen-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.3 g/mol
MDL Number: MFCD22392275
CAS Number: 1251734-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR