3,3-Dimethyl-6-nitroindolin-2-one 95%

Supplier: Ambeed
Warning

A352644-100MG A352644-1G A352644-250MG
76989-656EA 154.9 USD
76989-656 76989-658 76989-660
3,3-Dimethyl-6-nitroindolin-2-one 95%
3,3-Dimethyl-6-nitroindolin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MW: 206.2 g/mol
MDL Number: MFCD13191794
CAS Number: 100510-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR