(4-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A352424-1G
A352424-250MG
A352424-5G
77015-722EA
32.89
USD
77015-722
77015-724
77015-726
(4-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid 98%
(4-((4-(2-Methoxyethyl)phenoxy)methyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₆H₁₉BO₄ MW: 286.13 g/mol |
MDL Number:
MFCD07369748 CAS Number: 870779-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...