4-[4-(Hydroxymethyl)phenyl]pyridine 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
(4-Pyridin-4-ylphenyl)methanol, (4-(Pyridin-4-yl)phenyl)methanol
A352017-100MG
A352017-250MG
A352017-1G
77015-718EA
49.87
USD
77015-718
77015-720
77594-052
4-[4-(Hydroxymethyl)phenyl]pyridine 95%
4-[4-(Hydroxymethyl)phenyl]pyridine
Precaución: for Research Use Only
Formula:
C₁₂H₁₁NO MW: 185.222 g/mol |
MDL Number:
MFCD04114576 CAS Number: 217192-22-8 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...