Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-(methoxycarbonyl)-2-methyl-1,3-thiazole, Methyl 5-bromo-2-methylthiazole-4-carboxylate

A351756-1G A351756-250MG A351756-5G
76989-650EA 72.15 USD
76989-650 76989-652 76989-654
Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate 95%
Methyl 5-bromo-2-methyl-1,3-thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.086 g/mol
MDL Number: MFCD09265508
CAS Number: 899897-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR