N(α)-Cbz-L-histidine 98%

Supplier: Ambeed
Warning

Synonyms: Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH

A351518-1G A351518-5G A351518-25G
77003-256EA 10.61 USD
77003-256 77003-258 77672-772
N(α)-Cbz-L-histidine 98%
N(α)-Cbz-L-histidine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O₄
MW: 289.3 g/mol
MDL Number: MFCD00065960
CAS Number: 14997-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR