3-Cyano-3-methylazetidine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 3-Methylazetidine-3-carbonitrile hydrochloride, 3-Methyl-3-azetidinecarbonitrile hydrochloride

A351282-1G A351282-250MG A351282-5G A351282-100MG
76989-640EA 95.49 USD
76989-640 76989-642 77613-002 77684-038
3-Cyano-3-methylazetidine hydrochloride 95%
3-Cyano-3-methylazetidine hydrochloride

Precaución: For research use only.

Formula: C₅H₉ClN₂
MW: 132.59 g/mol
MDL Number: MFCD11849039
CAS Number: 936850-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR