1-Methyl-2-pyrrolecarboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: N-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrole-2-aldehyde, 1-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrol-2-carbaldehyde, 2-Formyl-1-methylpyrrole

A351000-100G A351000-10G A351000-25G A351000-5G
77003-246EA 129 USD
77003-246 77003-248 77003-250 77003-252
1-Methyl-2-pyrrolecarboxaldehyde 97%
1-Methyl-2-pyrrolecarboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00003087
CAS Number: 1192-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR