2-(4-(Trifluoromethyl)pyridin-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A350925-1G A350925-250MG
76989-636EA 107 USD
76989-636 76989-638
2-(4-(Trifluoromethyl)pyridin-2-yl)acetonitrile 97%
2-(4-(Trifluoromethyl)pyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂
MDL Number: MFCD09923755
CAS Number: 1000536-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR