Dimethyl-DL-glutamate hydrochloride 97%

Supplier: Ambeed
Warning

A350861-100G A350861-10G A350861-25G A350861-5G
77033-104EA 75 USD
77033-104 77033-106 77033-108 77033-110
Dimethyl-DL-glutamate hydrochloride 97%
Dimethyl-DL-glutamate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₄
MDL Number: MFCD00266895
CAS Number: 13515-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR