2-Amino-5-fluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

A349527-1G A349527-250MG
76989-598EA 372.38 USD
76989-598 76989-600
2-Amino-5-fluoroisonicotinic acid 97%
2-Amino-5-fluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD18250109
CAS Number: 1260671-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR