4-Phenoxypiperidine hydrochloride 98%

Supplier: Ambeed
Warning

A349106-1G A349106-250MG A349106-5G
77015-706EA 67 USD
77015-706 77015-708 77015-710
4-Phenoxypiperidine hydrochloride 98%
4-Phenoxypiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD02684106
CAS Number: 3413-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR