1-(4-Aminophenyl)cyclopentanecarbonitrile 97%

Supplier: Ambeed
Warning

A349043-1G A349043-250MG A349043-5G
77003-242EA 15 USD
77003-242 77003-244 77672-766
1-(4-Aminophenyl)cyclopentanecarbonitrile 97%
1-(4-Aminophenyl)cyclopentanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂
MDL Number: MFCD00807534
CAS Number: 115279-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR