3-Bromo-2-cyano-5-methoxypyridine 97%

Supplier: Ambeed
Warning

A348929-100MG A348929-1G A348929-250MG A348929-5G
77015-698EA 45 USD
77015-698 77015-700 77015-702 77015-704
3-Bromo-2-cyano-5-methoxypyridine 97%
3-Bromo-2-cyano-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂O
MDL Number: MFCD08062679
CAS Number: 717843-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR