5-Methyl-2-(trifluoromethoxy)bromobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methyl-1-(trifluoromethoxy)benzene

A348684-1G A348684-250MG
76989-578EA 116.7 USD
76989-578 76989-580
5-Methyl-2-(trifluoromethoxy)bromobenzene 95%
5-Methyl-2-(trifluoromethoxy)bromobenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
MDL Number: MFCD04972699
CAS Number: 887268-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR