Triapine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-AP, NSC 663249, 2-((3-Aminopyridin-2-yl)methylene)hydrazine-1-carbothioamide, 2-[(3-Aminopyridin-2-yl)methylene]hydrazinecarbothioamide, PAN-811, 3-Aminopyridine-2-Carboxyaldehyde Thiosemicarbazone
A348408-10MG
A348408-1MG
A348408-25MG
A348408-50MG
A348408-5MG
A348408-100MG
A348408-1G
A348408-250MG
76989-566EA
37.14
USD
76989-566
76989-568
76989-570
76989-572
76989-574
77684-030
77684-031
77684-032
Triapine 98%
Triapine
Precaución: For research use only.
Formula:
C₇H₉N₅S MW: 195.24 g/mol |
MDL Number:
MFCD00924747 CAS Number: 143621-35-6 UN: 2811 ADR: 6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...