Methyl 1-Cbz-5-Hydroxypiperidine-3-carboxylate 96%

Supplier: Ambeed
Warning

A348396-100MG A348396-1G A348396-250MG
77003-230EA 18.04 USD
77003-230 77003-232 77003-234
Methyl 1-Cbz-5-Hydroxypiperidine-3-carboxylate 96%
Methyl 1-Cbz-5-Hydroxypiperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₅
MW: 293.31 g/mol
MDL Number: MFCD11656768
CAS Number: 1095010-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR