3-Bromo-5-chloropyridine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-5-chloropicolinic acid
A348335-100G
A348335-10G
A348335-1G
A348335-250MG
A348335-25G
A348335-5G
77033-088EA
1277.33
USD
77033-088
77033-090
77033-092
77033-094
77033-096
77033-098
3-Bromo-5-chloropyridine-2-carboxylic acid 95%
3-Bromo-5-chloropyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₃BrClNO₂ MW: 236.46 g/mol |
MDL Number:
MFCD12827551 CAS Number: 1189513-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...