5-Chloro-3-fluoro-2-methoxypyridine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A348026-1G A348026-5G A348026-10G
77015-696EA 24 USD
77015-696 77684-028 77890-696
5-Chloro-3-fluoro-2-methoxypyridine 98%, Ambeed.Inc
5-Chloro-3-fluoro-2-methoxypyridine

Precaución: For research use only.

Formula: C₆H₅ClFNO
MDL Number: MFCD07375056
CAS Number: 886374-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR